Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chenodeoxycholic acid, Thermo Scientific Chemicals

Catalog No. AAJ6036414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6036414 25 g
AAJ6036406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6036414 Supplier Thermo Scientific Chemicals Supplier No. J6036414
Add to Cart

Bile acid that induces apoptosis through protein kinase C signaling pathways

Chenodeoxycholic acid is a bile acid that induces apoptosis through protein kinase C signaling pathways.Chenodeoxycholic acid is widely utilized in therapeutic applications. It is applied in medical therapy to dissolve gallstones. It is employed in the treatment of cerebrotendineous xanthomatosis. It is used to treat constipation and cerebrotendineous xanthomatosis. It acts as a urea receptor in supramolecular chemistry which can contain anions. It is a staining additive commonly used with ruthenium or organic photo-sensitizers in the preparation of staining solutions for dye solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water, dimethyl sulfoxide, ethanol and dimethylformamide.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 474-25-9
Molecular Formula C24H40O4
Molecular Weight (g/mol) 392.58
MDL Number MFCD00064142
InChI Key RUDATBOHQWOJDD-BSWAIDMHSA-N
Synonym chenodexycholic acid, chenodeoxycholate anion, chenodeoxycholate 1-, 3alpha, 7alpha-dihydroxy-5beta-cholanic acid, 3alpha,7alpha-dihydroxy-5beta-cholan-24-oate, 3alpha,7alpha-dihydroxy-5beta-cholan-24-oic acid anion, 4r-4-3r,5s,7r,8r,9s,10s,13r,14s,17r-10,13-dimethyl-3,7-bis oxidanyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1h-cyclopenta a phenanthren-17-yl pentanoate, 4r-4-3r,5s,7r,8r,9s,10s,13r,14s,17r-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1h-cyclopenta a phenanthren-17-yl pentanoate
PubChem CID 9548590
ChEBI CHEBI:36234
IUPAC Name (4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11aR)-4,7-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoic acid
SMILES [H][C@@]12CC[C@H]([C@H](C)CCC(O)=O)[C@@]1(C)CC[C@@]1([H])[C@@]2([H])[C@H](O)C[C@]2([H])C[C@H](O)CC[C@]12C

Specifications

Melting Point 164°C to 168°C
Color White
Quantity 25 g
Beilstein 3219887
Merck Index 14,2054
Solubility Information Soluble in water,dimethyl sulfoxide,ethanol and dimethylformamide.
Optical Rotation +12° (c=2 in Ethanol)
Formula Weight 392.57
Physical Form Crystalline Needles
Chemical Name or Material Chenodeoxycholic acid
Hazard Category H315-H319
Hazard Statement GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P501c
EINECSNumber 207-481-8
RTECSNumber FZ1980000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.