Learn More
Cayman Chemical Chenodeoxycholyl-cadaverine, Purity: ≥98%, CAS: 142970-49-8, 10 mg
SDP
Catalog No. 502945862
Encompass_Preferred
A polyamine bile amidate; has been used as a standard in the detection of chenodeoxycholyl-cadaverine in mammalian intestinal and fecal samples
Catalog No. 50-294-5862 Supplier Cayman Chemical Supplier No. 4089510 MG
A polyamine bile amidate; has been used as a standard in the detection of chenodeoxycholyl-cadaverine in mammalian intestinal and fecal samples