Learn More
Cayman Chemical Chenodeoycholc ACD 24Acyl 10mg
SDP
Catalog No. 502949094
Encompass_Preferred
A metabolite of CDCA; formed from CDCA by UGT1A3
Catalog No. 50-294-9094 Supplier Cayman Chemical Supplier No. 3322710 MG
A metabolite of CDCA; formed from CDCA by UGT1A3