Learn More
ATCC CheRiff_eGFP tet-on; spiking HEK cells; Embryonic Kidney; Human (Homo sapiens)
Catalog No. 502384924
Encompass
CheRiff_eGFP tet-on spiking HEK cells is a cell line exhibiting epithelial-like morphology that was isolated from embryonic kidney tissue. It has applications for synthetic electrophysiological systems, and is useful in testing channelrhodopsins and voltage indicators.
Catalog No. 50-238-4924 Supplier ATCC Supplier No. CRL3480
CheRiff_eGFP tet-on spiking HEK cells is a cell line exhibiting epithelial-like morphology that was isolated from embryonic kidney tissue. It has applications for synthetic electrophysiological systems, and is useful in testing channelrhodopsins and voltage indicators.