Learn More
Aviva Systems Biology Corp CHERP Antibody - middle region (ARP40643_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40643_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHERP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CHERP. Alternate names: DAN16, SCAF6, SRA1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-57566