Learn More
Bioworld CHES , 100 g
Supplier:  Bioworld 403300321
Encompass_Preferred
CHES2-[N-cyclohexylamino]ethane sulfonic acid A zwitterionic buffering agent useful at pH 8.6-10.0, pKa 9.49 (25°C) Technical Information:pKa (20°C): 9.55PH Value: 8.6 - 10.0
Catalog No. 50-255-814