Learn More
Mitegen CHES/ NaOH, 1M, pH9.5, 250 ml, CAS # 103-47-9
SDP
Catalog No. NC1251251
Encompass_Preferred
Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging.�Our stock solutions are intended for Research and Development Use Only.
Catalog No. NC1251251 Supplier Mitegen Supplier No. 1008158
Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging.�Our stock solutions are intended for Research and Development Use Only.