Learn More
Research Products International Corp Chest Freezer Rack for 96-Deep Well Plates, Rack Dimensions (Inches) 3 5/8 x 5 1/2 x 23 1/2
SDP
Catalog No. 501977782
Encompass
Fits micro-plates with dimensions of 5'' x 3-3/8'' x 1-5/8'' H
Catalog No. 50-197-7782 Supplier Research Products International Corp Supplier No. 247558
Fits micro-plates with dimensions of 5'' x 3-3/8'' x 1-5/8'' H