Learn More
Labrepco Inc CHEST FREEZER RACK FOR 15ML &
SDP
Catalog No. 509020216
Encompass_Preferred
Rack, Chest Freezer; Labrepco; For 15mL and 50mL tube boxes; With locking rod; Lock dimension: 23.625 in.(595mm); Rack height: 25.0625 in.(637mm); Box capacity: 5
Catalog No. 50-902-0216 Supplier Labrepco Inc Supplier No. CFLB5
Rack, Chest Freezer; Labrepco; For 15mL and 50mL tube boxes; With locking rod; Lock dimension: 23.625 in.(595mm); Rack height: 25.0625 in.(637mm); Box capacity: 5