Learn More
Thomas Scientific Vertical Freezer Rack for 2" box, 10 cap
SDP
Catalog No. 50196065
Encompass_Preferred
Stainless Steel Vertical Freezer Rack for Chest Style and LN2 Freezers for use with 2" or 3" Boxes
Catalog No. 50-196-065 Supplier Thomas Scientific Supplier No. 1201Y57
Stainless Steel Vertical Freezer Rack for Chest Style and LN2 Freezers for use with 2" or 3" Boxes