Learn More
Thomas Scientific Vertical Freezer Rack for 3" box, 8 cap
SDP
Catalog No. 50196074
Encompass_Preferred
Stainless Steel Vertical Freezer Rack for Chest Style and LN2 Freezers for use with 2" or 3" Boxes
Catalog No. 50-196-074 Supplier Thomas Scientific Supplier No. 1201Y67
Stainless Steel Vertical Freezer Rack for Chest Style and LN2 Freezers for use with 2" or 3" Boxes