Learn More
Alkali Scientific Alkali Scientific™ Chest Freezer Racks for 96-Well and 384-Well Microtiter Plates | 8 Shelves High | Rack Only
SDP
Catalog No. 503029377
Encompass_Preferred
For 96-Well and 384-Well Microtiter Plates Plate Capacity: 40 with lids 48 without lids The CFMP-8 chest freezer racks are designed for storage of 96-well & 384-well microtiter plates, are 8 shelves high for storage in an LN2 or chest freezer. These freezer racks are made out of a durable corrosion resistant stainless-steel material. Each rack compartment holds 5 plates with lids for a total of 40 or 6 plates without lids for a total of 48 plates per rack. Security lock devices are also available for each rack though sold separately. These racks do not come standard with 96-well or 384-well microtiter plates. *Rack Only Dimensions (DxWxH): 3 5/8 x 5 1/2 x 26 3/4 inches (92 x 140 x 679mm)Compartment Dimensions (h x d x w): 3 5/16 x 3 7/16 x 5 3/16 inch (83.5 x 88 x 131mm) *Lock device is ordered separately.
Catalog No. 50-302-9377 Supplier Alkali Scientific Supplier No. CFMP8
For 96-Well and 384-Well Microtiter Plates Plate Capacity: 40 with lids 48 without lids The CFMP-8 chest freezer racks are designed for storage of 96-well & 384-well microtiter plates, are 8 shelves high for storage in an LN2 or chest freezer. These freezer racks are made out of a durable corrosion resistant stainless-steel material. Each rack compartment holds 5 plates with lids for a total of 40 or 6 plates without lids for a total of 48 plates per rack. Security lock devices are also available for each rack though sold separately. These racks do not come standard with 96-well or 384-well microtiter plates. *Rack Only Dimensions (DxWxH): 3 5/8 x 5 1/2 x 26 3/4 inches (92 x 140 x 679mm)Compartment Dimensions (h x d x w): 3 5/16 x 3 7/16 x 5 3/16 inch (83.5 x 88 x 131mm) *Lock device is ordered separately.