Learn More
Alkali Scientific Alkali Scientific™ Chest Freezer Racks for 96-Well and 384-Well Microtiter Plates | 7 Shelves High | Rack Only
SDP
Catalog No. 503029486
Encompass_Preferred
For 96-Well and 384-Well Microtiter Plates Plate Capacity: 35 with lids 42 without lids The chest freezer racks are designed for storage of 96-well & 384-well microtiter plates, are 7 shelves high for storage in an LN2 or chest freezer. These freezer racks are made of a durable corrosion resistant stainless-steel material. Each rack compartment holds 5 plates with lids for a total of 35 or 6 plates without lids for a total of 42 plates per rack. Security lock devices are also available for each rack though sold separately. These racks do not come standard with 96-well or 384-well microtiter plates. *Rack Only Dimensions (DxWxH): 3 5/8 x 5 1/2 x 23 7/16 inches (92 x 140 x 595mm)Compartment Dimensions (h x d x w): 3 5/16 x 3 7/16 x 5 3/16 inch (83.5 x 88 x 131mm) *Lock device is ordered separately.
Catalog No. 50-302-9486 Supplier Alkali Scientific Supplier No. CFMP7
For 96-Well and 384-Well Microtiter Plates Plate Capacity: 35 with lids 42 without lids The chest freezer racks are designed for storage of 96-well & 384-well microtiter plates, are 7 shelves high for storage in an LN2 or chest freezer. These freezer racks are made of a durable corrosion resistant stainless-steel material. Each rack compartment holds 5 plates with lids for a total of 35 or 6 plates without lids for a total of 42 plates per rack. Security lock devices are also available for each rack though sold separately. These racks do not come standard with 96-well or 384-well microtiter plates. *Rack Only Dimensions (DxWxH): 3 5/8 x 5 1/2 x 23 7/16 inches (92 x 140 x 595mm)Compartment Dimensions (h x d x w): 3 5/16 x 3 7/16 x 5 3/16 inch (83.5 x 88 x 131mm) *Lock device is ordered separately.