Learn More
Alkali Scientific Alkali Scientific™ Chest Freezer Racks for 96-Well and 384-Well Microtiter Plates | 9 Shelves High | Rack Only
SDP
Catalog No. 503029362
Encompass_Preferred
  For 96-Well and 384-Well Microtiter Plates Plate Capacity: 45 with lids 54 without lids The chest freezer racks are designed for storage of 96-well & 384-well microtiter plates, are 9 shelves high for storage in an LN2 or chest freezer. These freezer racks are made of a durable corrosion resistant stainless-steel material. Each rack compartment holds 5 plates with lids for a total of 45 or 6 plates without lids for a total of 54 plates per rack. Security lock devices are also available for each rack though sold separately. These racks do not come standard with 96-well or 384-well microtiter plates. *Microtiter plates not included Dimensions (DxWxH): 3 5/8 x 5 1/2 x 30 1/16 inches (92 x 140 x 763mm)Compartment Dimensions (h x d x w): 3 5/16 x 3 7/16 x 5 3/16 inch (83.5 x 88 x 131mm) .*Lock device is ordered separately.
Catalog No. 50-302-9362 Supplier Alkali Scientific Supplier No. CFMP9
  For 96-Well and 384-Well Microtiter Plates Plate Capacity: 45 with lids 54 without lids The chest freezer racks are designed for storage of 96-well & 384-well microtiter plates, are 9 shelves high for storage in an LN2 or chest freezer. These freezer racks are made of a durable corrosion resistant stainless-steel material. Each rack compartment holds 5 plates with lids for a total of 45 or 6 plates without lids for a total of 54 plates per rack. Security lock devices are also available for each rack though sold separately. These racks do not come standard with 96-well or 384-well microtiter plates. *Microtiter plates not included Dimensions (DxWxH): 3 5/8 x 5 1/2 x 30 1/16 inches (92 x 140 x 763mm)Compartment Dimensions (h x d x w): 3 5/16 x 3 7/16 x 5 3/16 inch (83.5 x 88 x 131mm) .*Lock device is ordered separately.