Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chetomin | 1403-36-7 | MFCD28137713 | >99.0% | 710.87 g/mol | C31H30N6O6S4 | 1 MG
SDP

Catalog No. 5000241291
Encompass_Preferred

Chetomin is a natural-product small molecule reported to inhibit the Hsp90/HIF-1α pathway. Supplied as an off-white solid for laboratory research use, it is provided with recommended handling, solubility, and storage guidance for preparation of stock solutions.

  • Natural product inhibitor of Hsp90/HIF-1α pathway.
  • Off-white solid form, soluble in DMSO with warming and sonication.
  • Molecular formula C31H30N6O6S4; molecular weight 710.87 g/mol.
  • Recommended stock: DMSO, 5 mg/mL (7.03 mM).
  • Storage: powder -20°C; stock -80°C up to 6 months, -20°C up to 1 month.
  • For research use only; not intended for human or clinical use.

Catalog No. 50-002-41291 Supplier Medchemexpress LLC Supplier No. HY1075531MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chetomin is a natural-product small molecule reported to inhibit the Hsp90/HIF-1α pathway. Supplied as an off-white solid for laboratory research use, it is provided with recommended handling, solubility, and storage guidance for preparation of stock solutions.

  • Natural product inhibitor of Hsp90/HIF-1α pathway.
  • Off-white solid form, soluble in DMSO with warming and sonication.
  • Molecular formula C31H30N6O6S4; molecular weight 710.87 g/mol.
  • Recommended stock: DMSO, 5 mg/mL (7.03 mM).
  • Storage: powder -20°C; stock -80°C up to 6 months, -20°C up to 1 month.
  • For research use only; not intended for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.