Learn More
AdipoGen Chetomin
SDP
Catalog No. 502562081
Encompass_Preferred
Chemical. CAS 1403-36-7. Formula C31H30N6O6S4. MW 710.9. Isolated from Chaetomium sp. Antibiotic. Inhibitor of HIF-1 formation by disrupting the binding of p300 to both HIF-1alpha and HIF-2alpha. Histone H3K9 methyltransferases G9a and Suv39h inhibitor. Tumor growth inhibitor. Angiogenesis inhibitor. Potent immunosuppressor. Antibacterial and antifungal. Strong inducer of retinal pigment epithelium RPE.
Catalog No. 50-256-2081 Supplier AdipoGen Supplier No. BVT0161M005
Add to Cart
Add to Cart
Chemical. CAS 1403-36-7. Formula C31H30N6O6S4. MW 710.9. Isolated from Chaetomium sp. Antibiotic. Inhibitor of HIF-1 formation by disrupting the binding of p300 to both HIF-1alpha and HIF-2alpha. Histone H3K9 methyltransferases G9a and Suv39h inhibitor. Tumor growth inhibitor. Angiogenesis inhibitor. Potent immunosuppressor. Antibacterial and antifungal. Strong inducer of retinal pigment epithelium RPE.