Learn More
Aviva Systems Biology Corp CHFR Antibody - N-terminal region (ARP43250_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43250_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHFR. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CHFR. Alternate names: FLJ10796, FLJ33629, RNF116, RNF196ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58474