Learn More
enQuire Bioreagents Chromogranin A / CHGA Monoclonal [Clone CGA/413], Size: 100ug
Supplier:  enQuire Bioreagents 1113MSM9P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: CGA/413. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b kappa. Positive Control: PC12 cells. Adrenal gland, bowel, thyroid, pancreas, or pheochromocytoma. Cellular Staining: Finely granular cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human chromogranin A protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 68-75kDa.Chromosome: 14q32.12. Gene Symbol: CHGA. Gene ID: 1113. Ensemble: ENSG00000100604. Uniprot: NP_001266.
Catalog No. 50-188-4607