Learn More
enQuire Bioreagents Chromogranin A Recombinant Monoclonal [Clone CHGA/1815R], Size: 100ug
Supplier:  enQuire Bioreagents 1113RBM15P1
Encompass_Preferred
Product Type: Antibody. Application: WB, IHC. Conjugate/Tag: Unconjugated. Clone: CHGA/1815R. Clonality: Monoclonal. Host: Rabbit. Isotype: IgG, kappa. Positive Control: PC12 cells. Adrenal gland, bowel, thyroid, pancreas, or pheochromocytoma. Cellular Staining: Finely granular cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Also available without BSA & Azide. Concentration: 200 ug/ml. Purity: Protein A/G purified from Bioreactor concentrate. Immunogen: Recombinant full-length human CHGA protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 68-75kDa.Gene ID: 1113.
Catalog No. 50-188-4610