Learn More
Aviva Systems Biology Corp CHGA Antibody - C-terminal region (ARP41931_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41931_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHGA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CHGA. Alternate names: CGAApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58052