Learn More
Medchemexpress LLC CHI-KAT8i5 | 2839860-29-4 | 98.00% | 523.69 | 5 MG
SDP
Catalog No. 5000391028
Encompass_Preferred
CHI-KAT8i5 is a selective, orally active KAT8 inhibitor that induces cancer cell apoptosis and suppresses esophageal squamous cell carcinoma (ESCC) growth by targeting the KAT8/c-Myc signaling pathway. It demonstrates high specificity and anti-proliferative effects in vitro and in vivo.
  • Selective, orally active KAT8 inhibitor
  • High specificity; does not bind to other HAT family proteins
  • Induces cancer cell apoptosis
  • Suppresses esophageal squamous cell carcinoma (ESCC) growth
  • Targets KAT8/c-Myc signaling pathway
  • Demonstrates in vitro anti-proliferative effects against KYSE 30 and KYSE 150 cells
  • Represses NPM1 and PPAT transcript expression; decreases H4K16ace and c-Myc levels
  • Effective in vivo in suppressing ESCC tumor growth
  • Reduces Ki-67, c-Myc expression, and H4K16ace levels in vivo
Catalog No. 50-003-91028 Supplier Medchemexpress LLC Supplier No. HY1684285MG
CHI-KAT8i5 is a selective, orally active KAT8 inhibitor that induces cancer cell apoptosis and suppresses esophageal squamous cell carcinoma (ESCC) growth by targeting the KAT8/c-Myc signaling pathway. It demonstrates high specificity and anti-proliferative effects in vitro and in vivo.
  • Selective, orally active KAT8 inhibitor
  • High specificity; does not bind to other HAT family proteins
  • Induces cancer cell apoptosis
  • Suppresses esophageal squamous cell carcinoma (ESCC) growth
  • Targets KAT8/c-Myc signaling pathway
  • Demonstrates in vitro anti-proliferative effects against KYSE 30 and KYSE 150 cells
  • Represses NPM1 and PPAT transcript expression; decreases H4K16ace and c-Myc levels
  • Effective in vivo in suppressing ESCC tumor growth
  • Reduces Ki-67, c-Myc expression, and H4K16ace levels in vivo