Learn More
Medchemexpress LLC CHI-KAT8i5 | 98.0% | 523.69 | 25 MG
SDP
Catalog No. 5000391522
Encompass_Preferred
CHI-KAT8i5 is a selective and orally active KAT8 inhibitor that induces cancer cell apoptosis and suppresses esophageal squamous cell carcinoma (ESCC) growth by targeting the KAT8/c-Myc signaling pathway. It is a white to off-white solid.
  • Selective KAT8 inhibitor
  • Induces cancer cell apoptosis
  • Suppresses ESCC tumor growth
  • Soluble in DMSO at 100 mg/mL
  • Stable as powder for up to 3 years
Catalog No. 50-003-91522 Supplier Medchemexpress LLC Supplier No. 25MGHY168428
CHI-KAT8i5 is a selective and orally active KAT8 inhibitor that induces cancer cell apoptosis and suppresses esophageal squamous cell carcinoma (ESCC) growth by targeting the KAT8/c-Myc signaling pathway. It is a white to off-white solid.
  • Selective KAT8 inhibitor
  • Induces cancer cell apoptosis
  • Suppresses ESCC tumor growth
  • Soluble in DMSO at 100 mg/mL
  • Stable as powder for up to 3 years