Learn More
Aviva Systems Biology Corp CHI3L1 Antibody - middle region (ARP51929_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51929_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHI3L1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CHI3L1. Alternate names: DKFZp686N19119, FLJ38139, GP39, HC-gp39, HCGP-3P, YKL40, YYL-40, ASRT7, GP-39, CGP-39, YKL-40, hCGP-39ApplicationIF, WB. Species reactivity: Cow, Dog, Goat, Human, Pig, Rabbit, Rat
Catalog No. 50-014-60647