Learn More
Aviva Systems Biology Corp CHI3L2 Antibody - N-terminal region (ARP62213_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62213_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHI3L2. It was validated on Western Blot. . Alternate names: YKL-39, YKL39ApplicationWB. Species reactivity: Cow, Horse, Human, Pig, Rabbit
Catalog No. 50-287-612