Learn More
Aviva Systems Biology Corp CHIA Antibody - middle region (ARP42602_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42602_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHIA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CHIA. Alternate names: 2200003E03Rik, AMCase, CHIT2, DKFZp313J1722, RP5-1125M8.1, TSA1902, AMCASEApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-58281