Learn More
Active Motif ChIC/CUT&RUN Kit 24 rxns
SDP
Catalog No. NC2586634
Encompass
CUT&RUN (Cleavage Under Targets & Release Using Nuclease) is an epigenetic method used to investigate the genome-wide distribution of various chromatin-associated proteins and their modifications. CUT&RUN is a derivative of chromatin immunocleavage (ChIC). CUT&RUN is similar to chromatin immunoprecipitation (ChIP), in that it utilizes an antibody to target chromatin-associated marks and proteins, but requires less sample material and less sequencing depths than ChIP. CUT&RUN is compatible with 5,000 to 500,000 cells, and this assay kit is formatted with strip tubes for a more efficient workflow.
Catalog No. NC2586634 Supplier Active Motif Supplier No. 53180
CUT&RUN (Cleavage Under Targets & Release Using Nuclease) is an epigenetic method used to investigate the genome-wide distribution of various chromatin-associated proteins and their modifications. CUT&RUN is a derivative of chromatin immunocleavage (ChIC). CUT&RUN is similar to chromatin immunoprecipitation (ChIP), in that it utilizes an antibody to target chromatin-associated marks and proteins, but requires less sample material and less sequencing depths than ChIP. CUT&RUN is compatible with 5,000 to 500,000 cells, and this assay kit is formatted with strip tubes for a more efficient workflow.