Learn More
Aviva Systems Biology Corp CHIC1 Antibody - N-terminal region (ARP44893_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44893_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CHIC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CHIC1. Alternate names: RP11-108A15.1, BRX, DKFZp313P1931, DKFZp686F2342ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58930