Learn More
Aviva Systems Biology Corp CHIC2 Antibody - N-terminal region (ARP46856_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46856_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHIC2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human CHIC2. Alternate names: BTL, MGC21173ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53495