Learn More
MSC Chicago Hardware S-Hook, Low Carbon Steel, Electrogalvanized, 2-1/8 in
Catalog No. 5000536147
Encompass
This S-Hook is manufactured from low carbon steel and features an electrogalvanized zinc-plated finish for enhanced durability and corrosion resistance. With an overall length of 2-1/8 inches and a working load limit of 58 lb, it is suitable for a wide range of industrial and marine applications.
  • Made from low carbon steel.
  • Features an electrogalvanized zinc-plated finish.
  • Has an overall length of 2-1/8 in.
  • Offers a working load limit of 58 lb.
  • Wire diameter measures 0.220 in.
  • Opening in hooks is 5/16 in.
  • Inside diameter is 5/8 in.
  • Designed for marine and industrial applications.
Catalog No. 50-005-36147 Supplier MSC Supplier No. 06769889
This S-Hook is manufactured from low carbon steel and features an electrogalvanized zinc-plated finish for enhanced durability and corrosion resistance. With an overall length of 2-1/8 inches and a working load limit of 58 lb, it is suitable for a wide range of industrial and marine applications.
  • Made from low carbon steel.
  • Features an electrogalvanized zinc-plated finish.
  • Has an overall length of 2-1/8 in.
  • Offers a working load limit of 58 lb.
  • Wire diameter measures 0.220 in.
  • Opening in hooks is 5/16 in.
  • Inside diameter is 5/8 in.
  • Designed for marine and industrial applications.