Learn More
MSC Chicago Hardware S-Hook, Low Carbon Steel, Zinc Plated, 170 lb Capacity
Catalog No. 5000536148
Encompass
This S-hook, manufactured from low carbon steel, features a zinc plated finish for enhanced durability. Designed for a working load limit of 170 lb, it measures 3-1/2 in in overall length and is supplied with blunt ends. This versatile hook is ideal for various marine and industrial applications.
  • Constructed from low carbon steel wire.
  • Features a protective zinc plated finish.
  • Offers a working load limit of 170 lb.
  • Measures 3-1/2 in in overall length.
  • Equipped with 3/8 in diameter wire.
  • Has a 1 in inside hook diameter.
  • Includes a 1/2 in hook opening.
  • Supplied with blunt ends for safety.
  • Ideal for marine and industrial use.
Catalog No. 50-005-36148 Supplier MSC Supplier No. 06769913
This S-hook, manufactured from low carbon steel, features a zinc plated finish for enhanced durability. Designed for a working load limit of 170 lb, it measures 3-1/2 in in overall length and is supplied with blunt ends. This versatile hook is ideal for various marine and industrial applications.
  • Constructed from low carbon steel wire.
  • Features a protective zinc plated finish.
  • Offers a working load limit of 170 lb.
  • Measures 3-1/2 in in overall length.
  • Equipped with 3/8 in diameter wire.
  • Has a 1 in inside hook diameter.
  • Includes a 1/2 in hook opening.
  • Supplied with blunt ends for safety.
  • Ideal for marine and industrial use.