Learn More
MSC Chicago Protective Apparel Flame-Resistant Jacket, Small, 1 ea
Catalog No. 5000801958
Encompass
This flame-resistant jacket is made from a 9 oz., 100% cotton fabric that is treated with flame-retardant PROBAN® and cured with ammonia. If ignited, PROBAN will self-extinguish once the ignition source is removed. It is most effective as an outer layer but can be worn under aluminized clothing for high heat or molten splash exposure. This green cotton flame-resistant clothing is typically used in areas involving ferrous metals.
  • Made from 9 oz. 100% cotton fabric
  • Treated with flame-retardant PROBAN®
  • Self-extinguishes when ignition source is removed
  • Guaranteed to retain flame resistance for up to 25 industrial or 50 home launderings
  • Suitable for use in foundries, flame cutting, and welding
Catalog No. 50-008-01958 Supplier MSC Supplier No. 37780327
This flame-resistant jacket is made from a 9 oz., 100% cotton fabric that is treated with flame-retardant PROBAN® and cured with ammonia. If ignited, PROBAN will self-extinguish once the ignition source is removed. It is most effective as an outer layer but can be worn under aluminized clothing for high heat or molten splash exposure. This green cotton flame-resistant clothing is typically used in areas involving ferrous metals.
  • Made from 9 oz. 100% cotton fabric
  • Treated with flame-retardant PROBAN®
  • Self-extinguishes when ignition source is removed
  • Guaranteed to retain flame resistance for up to 25 industrial or 50 home launderings
  • Suitable for use in foundries, flame cutting, and welding