Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chicago Sky Blue 6B, Thermo Scientific™

Catalog No. AAA1424222
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1424222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1424222 Supplier Thermo Scientific Chemicals Supplier No. A1424222

C.I. 24410

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Chicago Sky Blue 6B is a counterstain for background autofluorescence in fluorescence and immunofluorescence histochemistry. It helps in the examination of blood plasma permeation.

Solubility
Soluble in water (1 mg/ml).

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2610-05-1
Molecular Formula C34H24N6Na4O16S4
Molecular Weight (g/mol) 992.79
MDL Number MFCD00004020
InChI Key UPKAWFACSJWKND-ZXFFUEEESA-J
Synonym C.I. 24410; Direct Blue 1
PubChem CID 44135407
IUPAC Name tetrasodium;4-amino-6-[[4-[4-[2-(8-amino-1-oxo-5,7-disulfonatonaphthalen-2-ylidene)hydrazinyl]-3-methoxyphenyl]-2-methoxyphenyl]hydrazinylidene]-5-oxonaphthalene-1,3-disulfonate
SMILES COC1=C(C=CC(=C1)C2=CC(=C(C=C2)NN=C3C=CC4=C(C3=O)C(=C(C=C4S(=O)(=O)[O-])S(=O)(=O)[O-])N)OC)NN=C5C=CC6=C(C5=O)C(=C(C=C6S(=O)(=O)[O-])S(=O)(=O)[O-])N.[Na+].[Na+].[Na+].[Na+]

Specifications

Solubility Information Soluble in water (1mg/ml).
Formula Weight 992.79
Quantity 100 g
Chemical Name or Material Chicago Sky Blue 6B
Hazard Category H350-H361d
Hazard Statement GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 220-026-8
RTECSNumber QJ6430000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.