Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chicago Sky Blue 6B, Tocris Bioscience™

Catalog No. 08461g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
08-461-G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 08-461-G Supplier R&D Systems Supplier No. 0846/1G
This item is not returnable. View return policy

Dye used as counterstain in histochemistry; also potent inhibitor of L-glutamate uptake into synaptic vesicles; also inhibits MIF

Potent inhibitor of L-glutamate uptake into synaptic vesicles. Allosterically inhibits macrophage migration inhibitory factor (MIF) (IC50 = 0.81 μM); inhibits MIF proinflammatory activity. Also dye used as counterstain in histochemistry.

Chemical Identifiers

CAS 2610-05-1
Molecular Formula C34H24N6Na4O16S4
Molecular Weight (g/mol) 992.79
InChI Key UPKAWFACSJWKND-ZXFFUEEESA-J
Synonym chicago blue 6b, pontamine sky blue 6bx, brillant benzo blue 6ba, tetrasodium 4-amino-6-2-4'-2-8-amino-1-hydroxy-5,7-disulfonatonaphthalen-2-yl diazen-1-yl-3,3'-dimethoxy-1,1'-biphenyl-4-yl diazen-1-yl-5-hydroxynaphthalene-1,3-disulfonate, tetrasodium 4-amino-6-e-2-4'-e-2-8-amino-1-hydroxy-5,7-disulfonatonaphthalen-2-yl diazen-1-yl-3,3'-dimethoxy-1,1'-biphenyl-4-yl diazen-1-yl-5-hydroxynaphthalene-1,3-disulfonate
PubChem CID 44135407
IUPAC Name tetrasodium;4-amino-6-[[4-[4-[2-(8-amino-1-oxo-5,7-disulfonatonaphthalen-2-ylidene)hydrazinyl]-3-methoxyphenyl]-2-methoxyphenyl]hydrazinylidene]-5-oxonaphthalene-1,3-disulfonate
SMILES COC1=C(C=CC(=C1)C2=CC(=C(C=C2)NN=C3C=CC4=C(C3=O)C(=C(C=C4S(=O)(=O)[O-])S(=O)(=O)[O-])N)OC)NN=C5C=CC6=C(C5=O)C(=C(C=C6S(=O)(=O)[O-])S(=O)(=O)[O-])N.[Na+].[Na+].[Na+].[Na+]

Specifications

Quantity 1 g
Formula Weight 992.79
Percent Purity >98%
Chemical Name or Material Chicago Sky Blue 6B
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.