Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chick Life Cycle Exploration Set
SCI_ED

Catalog No. s98475
Change view
Click to view available options
Grade:
K+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S98475 K+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S98475 Supplier Learning Resources, Inc Supplier No. LER2733
Only null left
Add to Cart
Add to Cart

Crack open set of 21 eggs and see the day-by-day development of a chick.

Features realistic illustrations of a developing chick in eggs 1 to 20, plastic 3-D chick inside the final egg.

  • Invites children to make hands-on connections to life science and life cycles
  • Reinforces concepts and related vocabulary with multilingual Activity Guide that includes definitions, chicken facts and a blackline master to label
  • Enables self-checking and easy organization with numbered label on each egg and numbered places in storage tray
  • Durable plastic eggs measure 2.75 in.L each; plastic tray measures 15L x 8.5 in.W

Order Info

Grades K to 4.

Specifications

For Use With (Application) Illustrates the developing chick in eggs 1-20, plastic 3D chick inside the final egg
Grade K+
Includes 21 Plastic eggs (realistic illustrations in eggs 1 to 20, Plastic 3D chick in final egg), Plastic storage tray, multilingual activity guide with diagram, definitions and facts
Product Type Life Cycle, Chick Exploration Set
Grade Level Elementary, High School
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.