Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ ChickCheck: The Genetics Behind Blue Chicken Eggs
SDPSCI_ED

Catalog No. S001288 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
25 Students (5 Groups)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S001288 25 Students (5 Groups)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001288 Supplier EDVOTEK™ Supplier No. 343
Add to Cart
Add to Cart

Students will investigate the genetic basis of chicken egg color using PCR to detect a viral insertion associated with blue egg color.

  • Explore the genetics of chicken egg color using hands-on PCR.
  • Detect a viral DNA insertion near the SLCO1B3 gene that causes blue eggs.
  • Compare PCR band patterns to identify egg-color phenotypes.
  • Use genetic results to select breeding pairs for homozygous blue egg-laying chickens.
  • For 25 students working in 5 groups.
  • Includes : Instructions, PCR EdvoBeads™ PLUS, LyphoPrimer™ Mix, 100 bp Ladder, Control DNA and Chicken DNA, TE Buffer, Universal DNA Buffer, SYBR™ Safe DNA Stain, FlashBlue™ DNA Stain, Ultra-Spec Agarose™, TBE Electrophoresis Buffer Powder, 0.2 mL PCR Tubes, Plastic Micropestles, Snap-top Microcentrifuge Tubes

Specifications

Grade Level High School, Higher Ed
Grade 9 to 12
Time Range Set Up: 30-45 Min., PCR: 2 Hours or Overnight, Electrophoresis: 45 Min.
Class Size 25 Students (5 Groups)
Product Type Investigating Selective Breeding Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.