Learn More
Immunoreagents Inc CHICKEN A-GOAT IGG BIO 1 MG
Catalog No. 50271970
Encompass_Preferred
Secondary Antibody; Chicken anti-goat IgG (H&L), Affinity Pure, Biotin Conjugate, Host: Chicken, Format: Liq, Label: Biotin, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-271-970 Supplier Immunoreagents Inc Supplier No. CKXGT003DBIO
Secondary Antibody; Chicken anti-goat IgG (H&L), Affinity Pure, Biotin Conjugate, Host: Chicken, Format: Liq, Label: Biotin, Applications: WB, ELISA, IHC/ICC