Learn More
Immunoreagents Inc CHICKEN A-GOAT IGG(H&L) AP 2MG
Catalog No. 50271767
Encompass_Preferred
Secondary Antibody; Chicken anti-goat IgG (H&L), Affinity Pure, min x w/Hu, Mu or Rb lgG/SP, Host: Chicken, Format: Liq, Label: None, Applications: WB, ELISA, IHC/ICC, IF, IP, FC
Catalog No. 50-271-767 Supplier Immunoreagents Inc Supplier No. CKXGT003E
Secondary Antibody; Chicken anti-goat IgG (H&L), Affinity Pure, min x w/Hu, Mu or Rb lgG/SP, Host: Chicken, Format: Liq, Label: None, Applications: WB, ELISA, IHC/ICC, IF, IP, FC