Learn More
Immunoreagents Inc CHICKEN A-GOAT IGG HRP 0.5MG
Catalog No. 50271414
Encompass_Preferred
Secondary Antibody; Chicken anti-goat IgG (H&L), Affinity Pure, min x w/Hu,Mu or RB IgG/SP, HRP Conjugate, Host: Chicken, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-271-414 Supplier Immunoreagents Inc Supplier No. CKXGT003EHRPX
Secondary Antibody; Chicken anti-goat IgG (H&L), Affinity Pure, min x w/Hu,Mu or RB IgG/SP, HRP Conjugate, Host: Chicken, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC