Learn More
Immunoreagents Inc CHICKEN A-GOAT IGG HRP 1 MG
Catalog No. 50271390
Encompass_Preferred
Secondary Antibody; Chicken anti-goat IgG (H&L), Affinity Pure, HRP Conjugate, Host: Chicken, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-271-390 Supplier Immunoreagents Inc Supplier No. CKXGT003DHRPX
Secondary Antibody; Chicken anti-goat IgG (H&L), Affinity Pure, HRP Conjugate, Host: Chicken, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC