Learn More
Immunoreagents Inc CHICKEN A-MOUSE IGG AP
Catalog No. 50271787
Encompass_Preferred
Secondary Antibody; Chicken anti-mouse IgG (H&L) - Affinity Pure, min x w/Hu or Rb IgG/Serum Proteins, Host: Chicken, Format: Liq, Label: None, Applications: WB, ELISA, IHC/ICC, IF, IP, FC
Catalog No. 50-271-787 Supplier Immunoreagents Inc Supplier No. CKXMU003E
Secondary Antibody; Chicken anti-mouse IgG (H&L) - Affinity Pure, min x w/Hu or Rb IgG/Serum Proteins, Host: Chicken, Format: Liq, Label: None, Applications: WB, ELISA, IHC/ICC, IF, IP, FC