Learn More
Immunoreagents Inc CHICKEN A-MOUSE IGG BIO
Catalog No. 50271541
Encompass_Preferred
Secondary Antibody; Chicken anti-mouse IgG (H&L) - Affinity Pure, min x w/Hu or Rb IgG/Serum Proteins, Biotin Conjugate, Host: Chicken, Format: Liq, Label: Biotin, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-271-541 Supplier Immunoreagents Inc Supplier No. CKXMU003EBIO
Secondary Antibody; Chicken anti-mouse IgG (H&L) - Affinity Pure, min x w/Hu or Rb IgG/Serum Proteins, Biotin Conjugate, Host: Chicken, Format: Liq, Label: Biotin, Applications: WB, ELISA, IHC/ICC