Learn More
Immunoreagents Inc CHICKEN A-MOUSE IGG DYLIGHT550
Catalog No. 50271544
Encompass_Preferred
Secondary Antibody; Chicken anti-mouse IgG (H&L) - Affinity Pure, min x w/Hu or Rb IgG/Serum Proteins, DyLight 550 Conjugate, Host: Chicken, Format: Lyo, Label: DyL550, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-271-544 Supplier Immunoreagents Inc Supplier No. CKXMU003E550NHSX
Secondary Antibody; Chicken anti-mouse IgG (H&L) - Affinity Pure, min x w/Hu or Rb IgG/Serum Proteins, DyLight 550 Conjugate, Host: Chicken, Format: Lyo, Label: DyL550, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS