Learn More
Immunoreagents Inc CHICKEN A-MOUSE IGG HRP
Supplier:  Immunoreagents Inc CKXMU003DHRPX
Encompass_Preferred
Secondary Antibody; Chicken anti-mouse IgG (H&L) - Affinity Pure, HRP Conjugate, Host: Chicken, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-272-012