Learn More
Immunoreagents Inc CHICKEN A-RABBIT IGG ALP
Supplier:  Immunoreagents Inc CKXRB003EALP
Encompass_Preferred
Secondary Antibody; Chicken anti-rabbit IgG (H&L) - Affinity Pure, min x w/Hu or Mu IgG, ALP Conjugate, Host: Chicken, Format: Liq, Label: ALP, Applications: WB, ELISA
Catalog No. 50-271-620