Learn More
Immunoreagents Inc CHICKEN A-RABBIT IGG AP
Catalog No. 50271619
Encompass_Preferred
Secondary Antibody; Chicken anti-rabbit IgG (H&L) - Affinity Pure, Host: Chicken, Format: Liq, Label: None, Applications: WB, ELISA, IHC/ICC, IF, IP, FC
Catalog No. 50-271-619 Supplier Immunoreagents Inc Supplier No. CKXRB003D
Secondary Antibody; Chicken anti-rabbit IgG (H&L) - Affinity Pure, Host: Chicken, Format: Liq, Label: None, Applications: WB, ELISA, IHC/ICC, IF, IP, FC