Learn More
Immunoreagents Inc CHICKEN A-RABBIT IGG BIO
Catalog No. 50271621
Encompass_Preferred
Secondary Antibody; Chicken anti-rabbit IgG (H&L) - Affinity Pure, min x w/Hu or Mu IgG, Biotin Conjugate, Host: Chicken, Format: Liq, Label: Biotin, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-271-621 Supplier Immunoreagents Inc Supplier No. CKXRB003EBIO
Secondary Antibody; Chicken anti-rabbit IgG (H&L) - Affinity Pure, min x w/Hu or Mu IgG, Biotin Conjugate, Host: Chicken, Format: Liq, Label: Biotin, Applications: WB, ELISA, IHC/ICC