Learn More
Immunoreagents Inc CHICKEN A-RABBIT IGG FITC
Catalog No. 50271178
Encompass_Preferred
Secondary Antibody; Chicken anti-rabbit IgG (H&L) - Affinity Pure, min x w/Hu or Mu IgG, FITC Conjugate, Host: Chicken, Format: Liq, Label: FITC, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-271-178 Supplier Immunoreagents Inc Supplier No. CKXRB003EFITC
Secondary Antibody; Chicken anti-rabbit IgG (H&L) - Affinity Pure, min x w/Hu or Mu IgG, FITC Conjugate, Host: Chicken, Format: Liq, Label: FITC, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS