Learn More
Immunoreagents Inc CHICKEN A-RABBIT IGG HRP
Supplier:  Immunoreagents Inc CKXRB003EHRPX
Encompass_Preferred
Secondary Antibody; Chicken anti-rabbit IgG (H&L) - Affinity Pure, min x w/Hu or Mu IgG, HRP Conjugate, Host: Chicken, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-271-622