Learn More
Immunoreagents Inc CHICKEN A-RAT IGG ALP
Catalog No. 50271651
Encompass_Preferred
Secondary Antibody; Chicken anti-rat IgG (H&L) - Affinity Pure, min x w/Hu or Rb IgG or Serum Proteins, ALP Conjugate, Host: Chicken, Format: Liq, Label: ALP, Applications: WB, ELISA
Catalog No. 50-271-651 Supplier Immunoreagents Inc Supplier No. CKXRT003EALP
Secondary Antibody; Chicken anti-rat IgG (H&L) - Affinity Pure, min x w/Hu or Rb IgG or Serum Proteins, ALP Conjugate, Host: Chicken, Format: Liq, Label: ALP, Applications: WB, ELISA