Learn More
Immunoreagents Inc CHICKEN A-RAT IGG HRP
Catalog No. 50271653
Encompass_Preferred
Secondary Antibody; Chicken anti-rat IgG (H&L) - Affinity Pure, min x w/Hu or Rb IgG or Serum Proteins, HRP Conjugate, Host: Chicken, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-271-653 Supplier Immunoreagents Inc Supplier No. CKXRT003EHRPX
Secondary Antibody; Chicken anti-rat IgG (H&L) - Affinity Pure, min x w/Hu or Rb IgG or Serum Proteins, HRP Conjugate, Host: Chicken, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC